Merck
  • Oligoarginine-Bearing Tandem Repeat Penetration-Accelerating Sequence Delivers Protein to Cytosol via Caveolae-Mediated Endocytosis.
Oligoarginine-Bearing Tandem Repeat Penetration-Accelerating Sequence Delivers Protein to Cytosol via Caveolae-Mediated Endocytosis.
Biomacromolecules (2019-03-21)
Akiko Okuda, Shinya Tahara, Hisaaki Hirose, Toshihide Takeuchi, Ikuhiko Nakase, Atsushi Ono, Masanori Takehashi, Seigo Tanaka, Shiroh Futaki
ABSTRACT
To facilitate the cytosolic delivery of larger molecules such as proteins, we developed a new cell-penetrating peptide sequence, named Pas2r12, consisting of a repeated Pas sequence (FFLIG-FFLIG) and d-dodeca-arginine (r12). This peptide significantly enhanced the cellular uptake and cytosolic release of enhanced green fluorescent protein and immunoglobulin G as cargos. We found that simply mixing Pas2r12 with cargos could generate cytosolic introducible forms. The cytosolic delivery of cargos by Pas2r12 was found to be an energy-requiring process, to rely on actin polymerization, and to be suppressed by caveolae-mediated endocytosis inhibitors (genistein and methyl-β-cyclodextrin) and small interfering RNA against caveolin-1. These results suggest that Pas2r12 enhances membrane penetration of cargos without the need for cross-linking and that caveolae-mediated endocytosis may be the route by which cytosolic delivery is enhanced.