Merck
  • Characterisation of cisplatin coordination sites in cellular Escherichia coli DNA-binding proteins by combined biphasic liquid chromatography and ESI tandem mass spectrometry.
Characterisation of cisplatin coordination sites in cellular Escherichia coli DNA-binding proteins by combined biphasic liquid chromatography and ESI tandem mass spectrometry.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2007-12-25)
Joanna Will, William S Sheldrick, Dirk Wolters
ABSTRACT
Combined multidimensional liquid chromatography and electrospray ionisation tandem mass spectrometry was employed to analyse platinated tryptic peptides from Escherichia coli cells treated with the anticancer drug cis-[PtCl2(NH3)2] at pH 7.0. Prerequisites for the LC/LC/MS/MS analysis of protein targets that are fulfilled by cisplatin are (a) that the original protein binding sites have a high kinetic stability over the range 2.3 < pH < 8.5, and (b) that the metal fragment remains coordinated to a significant number of b+ and y+ peptide ions under MS/MS fragmentation conditions. Matching the MS/MS spectra of the platinated tryptic peptides to sequences of proteins in the E. coli database enabled the identification of 31 protein targets for cisplatin. Whereas six of these are high-abundance enzymes and ribosomal proteins in E. coli cells, five low-abundance DNA-binding proteins were also identified as specific targets. These include the DNA mismatch repair protein mutS, the DNA helicase II (uvrD) and topoisomerase I (top1). Two efflux proteins (acrD, mdtA), the redox regulator thioredoxin 1 (thiO) and the external filament-like type-1 fimbrial protein A chain (fimA1) were also characterised as specific cisplatin-binding proteins. Kinetically favoured carboxylate (D, E) and hydroxy (S, T, Y) O atoms were identified as the Pt coordination sites in 18 proteins and methionyl S atoms in 9 proteins.