Merck
  • Paraxanthine displaces the binding of [3H]SCH 23390 from rat striatal membranes.
Paraxanthine displaces the binding of [3H]SCH 23390 from rat striatal membranes.
European journal of pharmacology (1990-04-25)
S Ferré, T Guix, J Sallés, A Badia, P Parra, F Jané, M Herrera-Marschitz, U Ungerstedt, M Casas
ABSTRACT
We present evidence showing that paraxanthine (1,7-dimethylxanthine), the main metabolite of caffeine in man, displaces the binding of [3H]SCH 23390, a radioligand which selectively labels dopamine D-1 receptors when used at low concentrations, from striatal membranes of the rat. The displacement was competitive and indicated the existence of two affinity states (Hill coefficient = 0.49; K(high) = 0.15 microM; K(low) = 95.9 microM, %R(high) = 32.4). When the stable GTP analog Gpp(NH)p was included, the displacement curve indicated the presence of only the low-affinity state (Hill coefficient = 1.16; Ki = 72.1 microM). However, paraxanthine did not displace the specific binding of [3H]spiperone. After injection of 30 mg/kg s.c. of caffeine, a maximum of 10 microM of paraxanthine was found in striatal homogenates, which could be sufficient to occupy dopamine D-1 receptors. Our results suggest that a dopaminergic action of paraxanthine could be involved in the behavioural stimulation produced by caffeine.
Sigma-Aldrich
1,7-Dimethylxanthine, ~98%, solid