- Lipid modification of proteins through sortase-catalyzed transpeptidation.
Lipid modification of proteins through sortase-catalyzed transpeptidation.
Journal of the American Chemical Society (2008-11-08)
John M Antos, Gwenn M Miller, Gijsbert M Grotenbreg, Hidde L Ploegh
PMID18989959
ABSTRACT
A general chemoenzymatic method for the site-specific attachment of lipids to protein substrates is described. Sortase A is used to append short lipid-modified oligoglycine peptides to the C terminus of protein substrates bearing a five amino acid sortase A recognition sequence (LPETG). We demonstrate the attachment of a range of hydrophobic modifications in excellent yield (60-90%), including a simple step for removing the sortase enzyme postreaction. Lipoproteins prepared using these procedures were subsequently shown to associate with mammalian cells in a lipid tail-dependent fashion and localized to the plasma membrane and endosomes.