Merck
  • Hydrophobic Amino Acid Content in Onions as Potential Fingerprints of Geographical Origin: The Case of Rossa da Inverno sel. Rojo Duro.
Hydrophobic Amino Acid Content in Onions as Potential Fingerprints of Geographical Origin: The Case of Rossa da Inverno sel. Rojo Duro.
Molecules (Basel, Switzerland) (2018-05-26)
Federica Ianni, Antonella Lisanti, Maura Marinozzi, Emidio Camaioni, Lucia Pucciarini, Andrea Massoli, Roccaldo Sardella, Luciano Concezzi, Benedetto Natalini
ABSTRACT
In this study, we were interested in comparing the amino acid profile in a specific variety of onion, Rossa da inverno sel. Rojo Duro, produced in two different Italian sites: the Cannara (Umbria region) and Imola (Emilia Romagna region) sites. Onions were cultivated in a comparable manner, mostly in terms of the mineral fertilization, seeding, and harvesting stages, as well as good weed control. Furthermore, in both regions, the plants were irrigated by the water sprinkler method and subjected to similar temperature and weather conditions. A further group of Cannara onions that were grown by micro-irrigation was also evaluated. After the extraction of the free amino acid mixture, an ion-pairing reversed-phase (IP-RP) HPLC method allowed for the separation and the evaporative light scattering detection of almost all the standard proteinogenic amino acids. However, only the peaks corresponding to leucine (Leu), phenylalanine (Phe), and tryptophan (Trp), were present in all the investigated samples and they were unaffected from the matrix interfering peaks. The use of the beeswarm/box plots revealed that the content of Leu and Phe were markedly influenced by the geographical origin of the onions (with *** p.