Merck
  • Dysfunction of HPV16-specific CD8+ T cells derived from oropharyngeal tumors is related to the expression of Tim-3 but not PD-1.
Dysfunction of HPV16-specific CD8+ T cells derived from oropharyngeal tumors is related to the expression of Tim-3 but not PD-1.
Oral oncology (2018-06-19)
Kamila Hladíková, Simona Partlová, Vladimír Koucký, Jan Bouček, Jean-Francois Fonteneau, Michal Zábrodský, Ruth Tachezy, Marek Grega, Radek Špíšek, Anna Fialová
ABSTRACT
Human papillomavirus (HPV) type 16 infection is one of the most important etiological agents of oropharyngeal squamous cell carcinoma. Patients with HPV-associated carcinomas of the head and neck were reported to have a better clinical outcome than patients with HPV-negative tumors. Because HPV16 E6 and E7 oncoproteins are highly immunogenic and constitutively expressed, HPV-specific T cell immunity may play the key role in improving the prognosis of these patients. Tumor-derived T cells were expanded in high levels of IL-2 and stimulated with HPV16 E6/E7 peptides in the presence or absence of anti-PD-1 monoclonal antibody nivolumab and soluble Tim-3. HPV16-specific tumor-infiltrating T cells were present in 73.1% of HPV-associated oropharyngeal tumors. HPV16 specific CD8+ TILs were able to produce IFNγ upon specific stimulation and predominantly expressed PD-1 but not Tim-3. Specific IFNγ production was further enhanced after a blockade of both PD-1 and Tim-3 pathways but not after a PD-1 blockade alone. Additionally, the specific stimulation of anti-HPV16 CD8+ T cells suppressed Tim-3 upregulation after the PD-1 blockade. Our data provide the rationale for combination cancer immunotherapy approaches, including the dual blockade of PD-1 and Tim-3 and, potentially, the use of HPV16-directed therapeutic vaccines.