Skip to Content
TW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xylometazoline hydrochloride
Synonym(s):
2-(4-
tert
-Butyl-2,6-dimethylbenzyl)-2-imidazoline hydrochloride, Xylometazoline hydrochloride
Empirical Formula (Hill Notation):
C
16
H
24
N
2
· HCl
CAS Number:
1218-35-5
Molecular Weight:
280.84
EC Number:
214-936-4
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (3)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
X6000
Xylometazoline hydrochloride,
Expand
View Pricing
1721002
Xylometazoline hydrochloride,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
X0100000
Xylometazoline hydrochloride,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP353
Xylometazoline hydrochloride,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing