Skip to Content
TW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Viresolve
®
Barrier Capsule
Products
Brand
Millipore (5)
Product Line
Surface Area cm²
Color
Membrane Material
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
VBMSPDKNB9
Viresolve
®
Barrier Capsule,
Process Development Kit
Expand
View Pricing
VBKG005
Viresolve
®
Barrier Capsule,
Retention of viruses, mycoplasma and bacteria from cell culture media
Expand
View Pricing
VBKG015
Viresolve
®
Barrier Capsule,
Retention of viruses, mycoplasma and bacteria from cell culture media
Expand
View Pricing
VBKG100HB1
Viresolve
®
Barrier Capsule,
Retention of viruses, mycoplasma and bacteria from cell culture media
Expand
View Pricing
VBKG050
Viresolve
®
Barrier Capsule,
Retention of viruses, mycoplasma and bacteria from cell culture media
Expand
View Pricing