Skip to Content
Merck
(R)-(+)-1,2-Dithiolane-3-pentanoic acid 97%

(R)-(+)-1,2-Dithiolane-3-pentanoic acid

(R)-(+)-1,2-Dithiolane-3-pentanoic acid 97%
Synonym(s):
(R)-6,8-Dithiooctanoic acid, (R)-6,8-Thioctic acid
Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing