Skip to Content
Merck
Loganic acid ≥95% (LC/MS-ELSD)

Loganic acid

Loganic acid ≥95% (LC/MS-ELSD)
Empirical Formula (Hill Notation):
C16H24O10
CAS Number:
Molecular Weight:
376.36
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing