Skip to Content
Merck
vibrant-m

Hydrocodone for peak identification

vibrant-m
Synonym(s):
Hydrocodone bitartrate salt hemi(pentahydrate)
Empirical Formula (Hill Notation):
C18H21NO3 · C4H6O6 · 2.5H2O
CAS Number:
Molecular Weight:
494.49

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1