![Etidronate disodium Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/109/127/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f/640/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f.png)
Etidronate disodium Related Compound A
![Etidronate disodium Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/109/127/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f/640/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f.png)
Synonym(s):
Sodium phosphite dibasic pentahydrate, di-Sodium hydrogen phosphite
Linear Formula:
Na2HPO3 · 5H2O
CAS Number:
Molecular Weight:
216.04
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing