Skip to Content
Merck
Carbenicillin disodium salt 89.0-100.5% anhydrous basis

Carbenicillin disodium salt

Carbenicillin disodium salt 89.0-100.5% anhydrous basis
Synonym(s):
α-Carboxybenzylpenicillin disodium salt
Empirical Formula (Hill Notation):
C17H16N2Na2O6S
CAS Number:
Molecular Weight:
422.36
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Application
Mode of Action
Special Grade
Technique