Anti-ZC3H4 antibody produced in rabbit
Synonym(s):
Anti-C19orf7, Anti-Kiaa1064, Anti-Zinc finger ccch-type containing 4
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1