Skip to Content
Merck
3-Methylpentane analytical standard

3-Methylpentane

3-Methylpentane analytical standard
Linear Formula:
(CH3CH2)2CHCH3
CAS Number:
Molecular Weight:
86.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing