
16-Doxyl-stearic acid methyl ester

Synonym(s):
2-Ethyl-2-(15-methoxy-15-oxopentadecyl)-4,4-dimethyl-3-oxazolidinyloxy, free radical
Empirical Formula (Hill Notation):
C23H44NO4
CAS Number:
Molecular Weight:
398.60
Products
Product Category
Brand
Formula Weight
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing