Product Name
1-(3-Pyridinylsulfonyl)-1H-benzotriazole 95%
Product Number
647330
Product Brand
aldrich
CAS Number
678173-43-8
Molecular Weight
260.27
Test | Specification |
---|---|
APPEARANCE | WHITE TO BROWN POWDER, CRYSTALS OR |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 47.7% - 53.8% |
NITROGEN 20.2% - 22.8% | |
SULFUR 11.5% - 13.1% | |
HIGH PRESSURE LIQUID | 94.0% (MINIMUM) |
APPROVED DECEMBER 9, 2005 BTR |