Product Name
2,4,6-Trimethylbenzyl mercaptan 98%
Product Number
428450
Product Brand
aldrich
CAS Number
21411-42-7
Molecular Weight
166.28
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Powder with Chunk(s)
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE