Product Name
5,5-Dimethylbarbituric acid ≥98.0%
Product Number
39566
Product Brand
ALDRICH
CAS Number
24448-94-0
Molecular Weight
156.14
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
TITRATION (T) NAOH 0.1M | 98.0 - 102.0 % |
MELTING POINT | 277 - 280 C |
SOLUBILITY (COLOR) | Colorless to Almost Colorless |
SOLUBILITY (TURBIDITY) | Clear to Hazy |
SOLUBILITY (METHOD) | 0.5G IN 10ML 5M SODIUM HYDROXIDE |
RESIDUE ON IGNITION | ≤ 0.10 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |