Skip to Content
Merck
All Photos(1)

Key Documents

COMH95E96275

Combi-Blocks, Inc.

Benzyl 2-(((cyclohexylmethyl)amino)methyl)-5-hydroxypiperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
360.5
MDL number:

Iupac Name

benzyl 2-(((cyclohexylmethyl)amino)methyl)-5-hydroxypiperidine-1-carboxylate

Inchi Code

1S/C21H32N2O3/c24-20-12-11-19(14-22-13-17-7-3-1-4-8-17)23(15-20)21(25)26-16-18-9-5-2-6-10-18/h2,5-6,9-10,17,19-20,22,24H,1,3-4,7-8,11-16H2

InChI key

JHSAVRZCPOJXRC-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HI-2445

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HI-2445


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service