CIAH9884FD79
ChemScene LLC (Preferred Partner)
Methyl 5-amino-2,3-dihydrobenzofuran-7-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
393.9+-42.0 C at 760 mmHg
Iupac Name
methyl 5-amino-2,3-dihydrobenzofuran-7-carboxylate
Inchi Code
1S/C10H11NO3/c1-13-10(12)8-5-7(11)4-6-2-3-14-9(6)8/h4-5H,2-3,11H2,1H3
InChI key
VHUHNEWQERKNBR-UHFFFAOYSA-N
storage temp.
4C, protect from light
Purity
96%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/1616110-67-8.html
Boiling point
393.9+-42.0 C at 760 mmHg
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0160732.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service