APO455825710
Apollo Scientific Ltd
1,2,3-Benzothiadiazole-5-carbonyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
93-96
Iupac Name
1,2,3-benzothiadiazole-5-carbonyl chloride
Inchi Code
1S/C7H3ClN2OS/c8-7(11)4-1-2-6-5(3-4)9-10-12-6/h1-3H
InChI key
VOSTUOLSYXVSND-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
95%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/123-benzothiadiazole-5-carbonyl-chloride
Boiling point
93-96
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/123-benzothiadiazole-5-carbonyl-chloride
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service