Skip to Content
Merck
All Photos(1)

Key Documents

729337

Sigma-Aldrich

tert-Butyldicyclohexylphosphonium tetrafluoroborate

97%

Synonym(s):

Dicyclohexyl-t-butylphosphonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H32BF4P
CAS Number:
Molecular Weight:
342.20
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

221-231 °C

functional group

phosphine

SMILES string

F[B-](F)(F)F.CC(C)(C)[PH+](C1CCCCC1)C2CCCCC2

InChI

1S/C16H31P.BF4/c1-16(2,3)17(14-10-6-4-7-11-14)15-12-8-5-9-13-15;2-1(3,4)5/h14-15H,4-13H2,1-3H3;/q;-1/p+1

InChI key

XOMLPCCNVCCMRG-UHFFFAOYSA-O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service