Merck
  • Inter-laboratory skin distribution study of 4-n-butyl resorcinol: The importance of liquid chromatography/mass spectrometry (HPLC-MS/MS) bioanalytical validation.
Inter-laboratory skin distribution study of 4-n-butyl resorcinol: The importance of liquid chromatography/mass spectrometry (HPLC-MS/MS) bioanalytical validation.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-07-05)
William Wargniez, Elsa Jungman, Simon Wilkinson, Nathalie Seyler, Sébastien Grégoire
ABSTRACT
In the present study, three laboratories independently compared percutaneous absorption and distribution of 4-n-butylresorcinol, using human skin from five donors. Each laboratory used the same protocol for percutaneous absorption studies but different LC-MS/MS analytical methods to quantify the test compound. All laboratories respected the mass balance criteria (i.e. 100±15%; average 96.5-102% of applied dose). Regarding usual inter-lab variability, good agreement was observed for all compartments with the greatest difference in the epidermis: 3.3 fold increase. The data obtained demonstrate that robustness of skin absorption data rely on properly validated analytical methods including sample extraction and LC-MS/MS method. It also includes clearly defined cutaneous absorption protocol for dose skin preparation, application, washing and tape stripping.
Product Description
Supelco
4-Butylresorcinol, analytical standard