- Soft, Peelable Organogels from Partially Hydrolyzed Poly(vinyl acetate) and Benzene-1,4-diboronic Acid: Applications to Clean Works of Art.
Soft, Peelable Organogels from Partially Hydrolyzed Poly(vinyl acetate) and Benzene-1,4-diboronic Acid: Applications to Clean Works of Art.
ACS applied materials & interfaces (2017-08-09)
Teresa T Duncan, Barbara H Berrie, Richard G Weiss
PMID28787129
ABSTRACT
We have developed soft, peelable organogels from 40% hydrolyzed poly(vinyl acetate) (40PVAc) and benzene-1,4-diboronic acid (BDBA). The organic liquids gelated include dimethyl sulfoxide, dimethylformamide, tetrahydrofuran, 2-ethoxyethanol, and methanol. The rheology of these soft materials can be tuned by altering the concentration of the polymer and/or crosslinker. Insights into the mechanisms leading to gelation were obtained from