Merck
  • Self-assembling covalent organic framework functionalized magnetic graphene hydrophilic biocomposites as an ultrasensitive matrix for N-linked glycopeptide recognition.
Self-assembling covalent organic framework functionalized magnetic graphene hydrophilic biocomposites as an ultrasensitive matrix for N-linked glycopeptide recognition.
Nanoscale (2017-07-18)
Jiaxi Wang, Jie Li, Mingxia Gao, Xiangmin Zhang
ABSTRACT
The development of additional functions and applications of covalent organic framework (COF)-derived materials still remains highly desired. In our work, a novel COF-functionalized magnetic graphene biocomposite (MagG@COF-5) was first developed as an ultrasensitive hydrophilic matrix via a facile self-assembly method for efficiently recognizing N-linked glycopeptides. By integrating the characteristics of the magnetic graphene and COF-5 layer, the MagG@COF-5 owns features of an outstanding magnetic response, a high specific area, strong hydrophilic properties and a unique size-exclusion effect. Accordingly, the MagG@COF-5 biocomposite showed excellent performance in N-linked glycopeptide analysis with a low detection limit (0.5 fmol μL
Sigma-Aldrich
Benzene-1,4-diboronic acid, ≥95.0%