Merck
  • Subtissue thermal sensing based on neodymium-doped LaF₃ nanoparticles.
Subtissue thermal sensing based on neodymium-doped LaF₃ nanoparticles.
ACS nano (2013-01-15)
Uéslen Rocha, Carlos Jacinto da Silva, Wagner Ferreira Silva, Ilde Guedes, Antonio Benayas, Laura Martínez Maestro, Mónica Acosta Elias, Enrico Bovero, Frank C J M van Veggel, José Antonio García Solé, Daniel Jaque
ABSTRACT
In this work, we report the multifunctional character of neodymium-doped LaF₃ core/shell nanoparticles. Because of the spectral overlap of the neodymium emission bands with the transparency windows of human tissues, these nanoparticles emerge as relevant subtissue optical probes. For neodymium contents optimizing the luminescence brightness of Nd³⁺:LaF₃ nanoparticles, subtissue penetration depths of several millimeters have been demonstrated. At the same time, it has been found that the infrared emission bands of Nd³⁺:LaF₃ nanoparticles show a remarkable thermal sensitivity, so that they can be advantageously used as luminescent nanothermometers for subtissue thermal sensing. This possibility has been demonstrated in this work: Nd³⁺:LaF₃ nanoparticles have been used to provide optical control over subtissue temperature in a single-beam plasmonic-mediated heating experiment. In this experiment, gold nanorods are used as nanoheaters while thermal reading is performed by the Nd³⁺:LaF₃ nanoparticles. The possibility of a real single-beam-controlled subtissue hyperthermia process is, therefore, pointed out.
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lanthanum, pieces
Pricing and availability is not currently available.
Sigma-Aldrich
Lanthanum(III) fluoride, anhydrous, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Pricing and availability is not currently available.