Key Documents
TMO140410
Nunc® EasyFill Cell Factory System
10 layer, polystyrene, surface area 6320 cm2, sterile, vented, case of 4 ea
Synonym(s):
closed cell system layered storage rack, closed cell system rack
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
material
polystyrene
sterility
sterile
feature
vented
packaging
case of 4 ea
manufacturer/tradename
Nunc 140410
surface area
6320 cm2
binding type
Nunclon Delta surface
General description
Maximize space utilization with the Thermo Scientific Nunc EasyFill Cell Factory system. This single-use system, consisting of 10 layers, can accommodate the same volume as 36 T175 flasks. The user-friendly linear multi-layer format simplifies cell culture and enables seamless scalability, making it suitable for a wide range of applications, from research to large-scale commercial use.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)