Skip to Content
Merck
All Photos(1)

Key Documents

P3615

Sigma-Aldrich

Prion Protein Fragment 106-126 Human

≥90% (HPLC), solid

Synonym(s):

PrP [106-126]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H138N26O24S2
CAS Number:
Molecular Weight:
1912.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCSC)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@@H](N)CCCCN)[C@@H](C)O)C(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)NCC(O)=O

InChI

1S/C80H138N26O24S2/c1-38(2)28-53(72(122)90-36-61(113)114)98-60(112)33-86-57(109)32-89-78(128)62(39(3)4)105-79(129)63(40(5)6)104-70(120)44(10)93-59(111)35-87-65(115)41(7)94-68(118)45(11)96-69(119)46(12)95-67(117)43(9)92-58(110)34-88-66(116)42(8)97-73(123)51(22-26-131-14)100-76(126)54(29-48-31-85-37-91-48)102-74(124)50(21-17-19-25-82)99-75(125)52(23-27-132-15)101-77(127)55(30-56(84)108)103-80(130)64(47(13)107)106-71(121)49(83)20-16-18-24-81/h31,37-47,49-55,62-64,107H,16-30,32-36,81-83H2,1-15H3,(H2,84,108)(H,85,91)(H,86,109)(H,87,115)(H,88,116)(H,89,128)(H,90,122)(H,92,110)(H,93,111)(H,94,118)(H,95,117)(H,96,119)(H,97,123)(H,98,112)(H,99,125)(H,100,126)(H,101,127)(H,102,124)(H,103,130)(H,104,120)(H,105,129)(H,106,121)(H,113,114)/t41-,42-,43-,44-,45-,46-,47+,49-,50-,51-,52-,53-,54-,55-,62-,63-,64-/m0/s1

InChI key

XPZWWTIIKSODDO-MBNDGZRNSA-N

Gene Information

human ... PRNP(5621)

Amino Acid Sequence

Lys-Thr-Asn-Met-Lys-His-Met-Ala-Gly-Ala-Ala-Ala-Ala-Gly-Ala-Val-Val-Gly-Gly-Leu-Gly

Application

Used as a model to investigate neurodegeneration in prion diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service