MilliporeSigma
Fragrance material review on cis-3-hexenyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-24)
A Lapczynski, D McGinty, L Jones, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of cis-3-hexenyl salicylate when used as a fragrance ingredient is presented.