Merck
  • Chiral resolutions of (9-anthryl)methoxyacetic acid and (9-anthryl)hydroxyacetic acid by capillary electrophoresis.
Chiral resolutions of (9-anthryl)methoxyacetic acid and (9-anthryl)hydroxyacetic acid by capillary electrophoresis.
Chirality (2007-10-26)
Bao-Lin Li, Zhen-Guo Zhang, Li-Li Du, Wei Wang
ABSTRACT
The resolutions of (9-anthryl)methoxyacetic acid (9AMAA) and (9-anthryl)hydroxyacetic acid (9AHAA) were performed by capillary electrophoresis using hydroxypropyl-beta-cyclodextrin (HP-beta-CD) as a chiral selector. Various factors affecting migration time and resolutions of these compounds were investigated with a run voltage of 20 kV, column temperature 20 degrees C and 20 mM Tris-H(3)PO(4) buffer (pH 6.5) containing 5 mM HP-beta-CD for 9AMAA, or 10 mM HP-beta-CD for 9AHAA, (+/-)-9AMAA and (+/-)-9AHAA were successfully separated at Rs 3.27 and 1.92, respectively.
Sigma-Aldrich
Methoxyacetic acid, 98%