V001655
Fast Blue B Salt
Synonym(s):
o-Dianisidine bis(diazotized) zinc double salt, Azoic Diazo No. 48, DBB, Diazo Blue B, Naphthanil Diazo Blue B
About This Item
Recommended Products
mp
>300 °C (lit.)
SMILES string
[Cl-].[Cl-].Cl[Zn]Cl.COc1cc(ccc1[N+]#N)-c2ccc([N+]#N)c(OC)c2
InChI
1S/C14H12N4O2.4ClH.Zn/c1-19-13-7-9(3-5-11(13)17-15)10-4-6-12(18-16)14(8-10)20-2;;;;;/h3-8H,1-2H3;4*1H;/q+2;;;;;+2/p-4
InChI key
GPPKNJIWDULNQH-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B - Self-react. C
Storage Class Code
5.2 - Organic peroxides and self-reacting hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service