Skip to Content
Merck
All Photos(1)

Documents

64808

Supelco

Extraction Chamber

Synonym(s):

Extraction Chamber for Demountable Liquid/Liquid Extractor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

packaging

pkg of 1

application(s)

food and beverages

compatibility

for use with Liquid/Liquid Extractor-Concentrator Kit

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service