4S8743
EPA 610 Polynuclear Aromatic Hydrocarbons Mixture
certified reference material, in methanol: methylene chloride (1:1) (varied)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Agency
EPA 610,IP-7,TO-13
NIOSH 5506,5515
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
ampule of 1 mL
concentration
in methanol: methylene chloride (1:1) (varied)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
multi-component solution
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Recommended products
Legal Information
Analyte
Signal Word
Danger
Hazard Statements
Precautionary Statements
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service