SAB56000302
Anti-CD13 Rabbit Monoclonal Antibody
clone RM403, affinity purified immunoglobulin
Synonym(s):
Aminopeptidase N, Myeloid plasma membrane glycoprotein CD13
About This Item
Recommended Products
antibody form
affinity purified immunoglobulin
Quality Level
antibody product type
primary antibodies
clone
RM403, monoclonal
recombinant monoclonal
species reactivity
human
technique(s)
immunohistochemistry: 1:100- 1:200
western blot: 1:1000-1:2000
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ANPEP(290)
Specificity
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service