Skip to Content
Merck
All Photos(4)

Key Documents

SAB1404487

Sigma-Aldrich

Monoclonal Anti-TPMT antibody produced in mouse

clone 1D4, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~53.06 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPMT(7172)

General description

This gene encodes the enzyme that metabolizes thiopurine drugs via S-adenosyl-L-methionine as the S-methyl donor and S-adenosyl-L-homocysteine as a byproduct. Thiopurine drugs such as 6-mercaptopurine are used as chemotherapeutic agents. Genetic polymorphisms that affect this enzymatic activity are correlated with variations in sensitivity and toxicity to such drugs within individuals. A pseudogene for this locus is located on chromosome 18q. (provided by RefSeq)

Immunogen

TPMT (AAH05339, 1 a.a. ~ 245 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDGTRTSLDIEEYSDTEVQKNQVLTLEEWQDKWVNGKTAFHQEQGHQLLKKHLDTFLKGKSGLRVFFPLCGKAVEMKWFADRGHSVVGVEISELGIQEFFTEQNLSYSEEPITEIPGTKVFKSSSGNISLYCCSIFDLPRTNIGKFDMIWDRGALVAINPGDRKCYADTMFSLLGKKFQYLLCVLSYDPTKHPGPPFYVPHAEIERLFGKICNIRRLEKVDAFEERHKSWGIDCLFEKLYLLTEK

Application

Monoclonal Anti-TPMT antibody produced in mouse is suitable for capture ELISA, indirect ELISA, indirect immunofluorescence and western blot analysis.

Biochem/physiol Actions

TPMT (Thiopurine S-methyltransferase) mainly mediates different catalytic S-methylation reaction such as 6-mercaptopurine and 6-thioguanine. It also participates in the metabolism of some drugs such as thiopurine drugs, which is largely influenced by TPMT polymorphisms. Deficiency in TPMT gene causes an autosomal recessive disorder, childhood acute lymphoblastic leukemia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H L McLeod et al.
Leukemia, 14(4), 567-572 (2000-04-14)
Thiopurine methyltransferase (TPMT) catalyses the S-methylation of thiopurines, including 6-mercaptopurine and 6-thioguanine. TPMT activity exhibits genetic polymorphism, with about 1/300 inheriting TPMT deficiency as an autosomal recessive trait. If treated with standard doses of thiopurines, TPMTdeficient patients accumulate excessive thioguanine
Hyun-Young Kim et al.
Pharmacogenetics and genomics, 25(3), 143-146 (2015-01-08)
Thiopurine S-methyltransferase (TPMT) is a cytoplasmic enzyme involved in the metabolism of thiopurine drugs and its activity is largely influenced by polymorphisms of the TPMT gene. To date, more than 35 TPMT variants are known to be associated with reduced
Hamza Ben Zeglam et al.
The Libyan journal of medicine, 10, 27053-27053 (2015-03-31)
Thiopurine S-methyltransferase (TPMT) is a cytosolic enzyme that catalyses the S-methylation of 6-mercaptopurine and azathioprine. Low activity phenotypes are correlated with polymorphism in the TPMT gene. Patients with low or undetectable TMPT activity could develop severe myelosuppression when they are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service