Skip to Content
Merck
All Photos(1)

Key Documents

N1135

Sigma-Aldrich

Ninhydrin Reagent

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352100

SMILES string

OC1(O)C(=O)c2ccccc2C1=O

Looking for similar products? Visit Product Comparison Guide

Application

For use in automated amino acid analysis

Packaging

Our stability testing indicates that DMSO and methyl cellosolve-based ninhydrin reagents deteriorate with time. Therefore, Sigma supplies this reagent as a set. The separate packages assure a quality reagent with long shelf life. The in-lab reconstitution requires a few extra minutes, but ensures excellent reagent performance.
Packaged under Argon

Kit Components Only

Product No.
Description

  • DMSO:4M lithium acetate buffer, pH5.20 (3:1) 950 mL

  • hydrindantin dihydrate 660 mg

  • ninhydrin 19 g

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
S Moore
The Journal of biological chemistry, 243(23), 6281-6283 (1968-12-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service