Skip to Content
Merck
All Photos(1)

Documents

M0750

Sigma-Aldrich

D-(+)-Malic acid

≥97.0%

Synonym(s):

(R)-(+)-2-Hydroxysuccinic acid, D-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723540
EC Number:
MDL number:
UNSPSC Code:
12352000

Assay

≥97.0%

mp

98-102 °C (lit.)

SMILES string

O[C@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m1/s1

InChI key

BJEPYKJPYRNKOW-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene Llorente-Folch et al.
Journal of neurochemistry, 124(3), 347-362 (2012-12-12)
The mitochondrial transporter of aspartate-glutamate Aralar/AGC1 is a regulatory component of the malate-aspartate shuttle. Aralar deficiency in mouse and human causes a shutdown of brain shuttle activity and global cerebral hypomyelination. A lack of neurofilament-labeled processes is detected in the
Narendra Kumar Sharma et al.
Journal of proteomics, 79, 277-298 (2013-01-15)
Hypoxia is one of the major stressors at high altitude. Exposure to hypobaric hypoxia induces several adverse consequences to the structural and functional integrity of brain. In an attempt to understand the proteome modulation, we used 2-DE coupled with MALDI-TOF/TOF
Duoduo Chen et al.
Molecular microbiology, 87(6), 1237-1244 (2013-01-26)
Nicotine is an environmental toxicant in tobacco waste, imposing a serious hazard for human health. Some bacteria including Pseudomonas spp. strains are able to metabolize nicotine to non-toxic compounds. The pyrrolidine pathway of nicotine degradation in Pseudomonas putida S16 has
Bhavana Agrawal et al.
The Journal of biological chemistry, 288(10), 7351-7362 (2013-01-17)
Mitochondria are important targets of metal toxicity and are also vital for maintaining metal homeostasis. Here, we examined the potential role of mitochondria in homeostasis of nickel in the roots of nickel hyperaccumulator plant Alyssum murale. We evaluated the biochemical
Frank Norbert Gellerich et al.
IUBMB life, 65(3), 180-190 (2013-02-13)
This review focuses on problems of the intracellular regulation of mitochondrial function in the brain via the (i) supply of mitochondria with ADP by means of ADP shuttles and channels and (ii) the Ca(2+) control of mitochondrial substrate supply. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service