Skip to Content
Merck
All Photos(1)

Key Documents

EHU093681

Sigma-Aldrich

MISSION® esiRNA

targeting human BLOC1S5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCGTTTAGCCTCTGCTTCAAGCTTAGCAATATATTCAGTGGCACTCTTATATCAGAAGAAAGAAGTTTCTACTGGCATTCTGATTGGATATTTAAAGAAGGGTGCCACATTTTTCCAGCTTTAAGTGCCTATATATGATTATTTGAATGAAGAGGAGTAGGAAGGAAAATGGAATCAATATACTCTTTGTTAGTCAGAATTGATCATCTTTTCCATTGATTAGCTCAGAGAACTGTAGGTATAGACTTCTTAGAAAGATAATTTCATTCATTTCTATACAGGGTTAACAATAGTTTATTATTGTGGCTTAATGGGAAGGTATAGAAGCTACCTGTCATAGCCTCCTTGGCTGTTGGAAGGATGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilhelmina C M Duivenvoorden et al.
Oncology letters, 13(5), 3624-3630 (2017-05-20)
Endoplasmic reticulum (ER) protein ERp46 is a member of the protein disulfide isomerase family of oxidoreductases, which facilitates the reduction of disulfides in proteins and their folding. Accumulation of misfolded proteins has been implicated in cancer. The objectives of the
L Wang et al.
Oncogene, 34(36), 4735-4745 (2014-12-17)
Castration-resistant prostate cancer (CRPC) continues to be a major clinical problem and the mechanisms behind it remain unclear. Thioredoxin domain-containing protein 5 (TXNDC5) is involved in protein folding and chaperone activity, and its overexpression has been reported in multiple malignancies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service