Skip to Content
Merck
All Photos(1)

Documents

CLS430499

Corning® cap inserts for cryogenic vials

assorted colors, 100 each of white, blue, red, green, and yellow

Synonym(s):

cryovials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121704
NACRES:
NB.13

sterility

non-sterile

manufacturer/tradename

Corning 430499

color

assorted colors, 100 each of white, blue, red, green, and yellow

Looking for similar products? Visit Product Comparison Guide

General description

  • Cap inserts provide color coding for easy sample identification
  • Inserts are packaged in resealable bags
  • Cap inserts fit all Corning cryogenic vials except Corning No. 430289

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service