Skip to Content
Merck
All Photos(1)

Documents

2D12

91051509, Reactivity: Anti-Yellow Fever Virus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-Yellow Fever Virus

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a, kappa

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse P3X63Ag8.653 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against the envelope protein of the wild type and vaccine strains of yellow fever virus. It does not cross react with other flaviviruses.

Immunogen

Yellow fever virus, strain 17D

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 0.05mM 2-mercaptoethanol (2ME) + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service