Skip to Content
Merck
All Photos(1)

Documents

46111

Supelco

2-Methoxy-5-methylaniline

analytical standard

Synonym(s):

5-Methyl-o-anisidine, 6-Methoxy-m-toluidine, p-Cresidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
637071
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.7 (vs air)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

235 °C (lit.)

mp

50-52 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COc1ccc(C)cc1N

InChI

1S/C8H11NO/c1-6-3-4-8(10-2)7(9)5-6/h3-5H,9H2,1-2H3

InChI key

WXWCDTXEKCVRRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxy-5-methylaniline is grouped under the class of aromatic amines (AAs), which are extensively utilized as intermediates in the synthesis of rubbers, plastics, dyes, etc. AAs are deemed as environmental pollutants as well as human carcinogens.

Application

2-Methoxy-5-methylaniline may be used as an analytical standard for the determination of the analyte in wastewater, textiles, and commercial hair dye samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rama Jeevan Ganesh Jeevan et al.
Electrophoresis, 23(4), 584-590 (2002-03-01)
Analysis of carcinogenic substances is a high-priority area. Carcinogenic arylamines draw the analyst's attention because dyes and pigments are in production and used in large volumes. Identification of carcinogenic isomers of arylamines employing micellar electrokinetic chromatography (MEKC), a mode of
P Sutthivaiyakit et al.
Analytical and bioanalytical chemistry, 381(1), 268-276 (2005-02-08)
A confirmation method for the determination of 18 aromatic amines originating from azo dyes after reductive cleavage was developed. The method is based on the use of high-performance liquid chromatography/tandem mass spectrometry with atmospheric-pressure chemical ionization. For the identification of
Determination of some banned aromatic amines in waste water using micellar liquid chromatography
Mourya SK, et al.
Analytical Methods : Advancing Methods and Applications, 3(9), 2032-2040 (2011)
T L Horn et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45(1), 55-63 (2006-09-13)
A six-month study was conducted in p53(+/-) mice to evaluate the possible oncogenicity of resveratrol (3,5,4'-trihydroxy-trans-stilbene), a cancer chemopreventive agent present in grapes and other foods. p53(+/-) mice (25/sex/group) received daily gavage exposure to vehicle only (negative control), resveratrol doses
Janet M Petruska et al.
Toxicologic pathology, 30(6), 696-704 (2003-01-07)
The carcinogenic potential of chlorpromazine hydrochloride, a psychotropic agent, was assessed in the p53 heterozygous mouse assay. In a 4-week dose range finding study in p53 wild-type mice, doses of 20,40, 60, and 80 mg/kg were poorly tolerated because of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service