Skip to Content
Merck
All Photos(1)

Key Documents

06629

Supelco

3′-Aminoacetophenone

puriss. p.a., standard for GC, ≥99.5% (GC)

Synonym(s):

3-Acetylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4COCH3
CAS Number:
Molecular Weight:
135.16
Beilstein:
386009
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

standard for GC
puriss. p.a.

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

289-290 °C (lit.)

mp

94-98 °C (lit.)
94-98 °C

application(s)

environmental

SMILES string

CC(=O)c1cccc(N)c1

InChI

1S/C8H9NO/c1-6(10)7-3-2-4-8(9)5-7/h2-5H,9H2,1H3

InChI key

CKQHAYFOPRIUOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service