TANK00200
Storage and Distribution System
200 L storage tank (no relay) for AFS® systems, For optimal storage of Type 2 or Type 3 pure water
Synonym(s):
200 L Storage Tank (no relay)
About This Item
Recommended Products
material
polyethylene reservoir
packaging
pkg of 1 unit
parameter
≤40 L/min distribution flow rate
250 kg operating weight (551.2 lb)
H × W × D
102 cm (40.2 in.) × 60 cm (23.6 in.) × 70 cm (27.6 in.)
capacity
200 L
volume
200 L
200000 mL
compatibility
for use with AFS®
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Polyethylene (PE) reservoir allows for the minimal release of organics.
- A tank vent filter protects water from particulates, bacteria, and dissolved CO2.
- An aseptic overflow function maintains water quality by avoiding retro-contamination from the drain.
- The conical reservoir base allows complete draining, and facilitates rinsing during sanitization, while the smooth interior limits biofilm formation.
- Hermetically sealed cover prevents unfiltered air from entering.
- Optional ASM (Automatic Sanitization Module) prevents bacterial growth and biofilm build-up inside the reservoir during storage.
- Distribution pump is controlled directly by the water system unit and the distribution flow rate of up to 40 L/min.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service