MABX4043
Anti- human myeloperoxidase (MPO) clone 43
clone 43, from mouse
Synonym(s):
Myeloperoxidase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
General description
Antibody recognizes Myeloperoxidase. Monitoring of level of Myeloperoxidase (MPO) in plasma can be useful to assist in the diagnosis critical cardiac conditions such as a Myocardial Infarction and also as a prognostic marker for other Cardiovascular Diseases (CVD s)
Application
Confirmed matched pairing. Capture Antibody MABX4187-1KC; Detector Antibody MABX4043-1KC
Detect MPO using this mouse monoclonal Anti- human myeloperoxidase (MPO) clone 43 Antibody, Cat. No. MABX4043-1KC, validated for use in and Western Blotting and ELISA.
Research Category
Signaling
Signaling
Physical form
Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing PBS (pH 7.4) with 0.09% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service