Skip to Content
Merck
All Photos(1)

Documents

CMTI-1

Sigma-Aldrich

EmbryoMax Embryonic Stem Cell Line - Strain 129/SVEV, passage 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

description

Cell line type: Embryonic Stem Cells
Mouse Embryonic Stem Cells

technique(s)

cell culture | stem cell: suitable

General description

Embryonic Stem Cell Lines derived from murine 129/SVEV selected for high targeting efficiency and proven germ line transmission. Cells are supplied as 2.5 x 106 ES cells per vial (approximately 2.5 population doublings per passage, shipped frozen on dry ice) and are supplied at passage 11.

Packaging

2.5x106 ea

Storage and Stability

Place vials in the vapor phase of liquid nitrogen storage immediately upon receipt until it is convenient to proceed to subculture.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service