Skip to Content
Merck
All Photos(1)

Documents

AG60

Sigma-Aldrich

Prostatic Acid Phosphatase

Synonym(s):

PAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
eCl@ss:
32160405
NACRES:
NA.26

biological source

human

Quality Level

Assay

>95% (SDS-PAGE)

form

liquid

specific activity

864 U/mg

manufacturer/tradename

Chemicon®

concentration

3.28 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ACPP(55)

General description

Product Source: Human seminal fluid. Tested negative for HIV I/II, HIV-1 antigen, and HCV antibodies and Hepatitis B surface antigen.
Purified HUMAN PROSTATIC ACID PHOSPHATASE (PAP) Protien isolated from human seminal Fluid.

NOTE:

This product exhibits phosphatase activity in acid conditions in the presence of chromogenic phosphate substrates.

Physical form

Purified protein. Lyophilized from 0.2um filtered solution with 0.02mM Ammonium Bicarbonate.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bonnie Taylor-Blake et al.
PloS one, 5(1), e8674-e8674 (2010-01-20)
Thiamine monophosphatase (TMPase, also known as Fluoride-resistant acid phosphatase or FRAP) is a classic histochemical marker of small- to medium-diameter dorsal root ganglia (DRG) neurons and has primarily been studied in the rat. Previously, we found that TMPase was molecularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service