Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14748

Sigma-Aldrich

Tetramethylammonium hydroxide

(25% solution in water) for synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107

Pricing and availability is not currently available.

vapor pressure

23.33 hPa ( 20 °C)

Quality Level

pH

>13 (20 °C in H2O)

bp

110 °C/1013 hPa

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): 24.0 - 27.0 %
Density (d 20 °C/ 4 °C): 1.010 - 1.030

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service