Skip to Content
Merck
All Photos(1)

Key Documents

8.04521

Sigma-Aldrich

Hexafluoroacetone sesquihydrate

for synthesis

Synonym(s):

Hexafluoroacetone sesquihydrate, Perfluoroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3F6O · 1.5H2O
CAS Number:
Molecular Weight:
193.04
UNSPSC Code:
12352115
EC Index Number:
211-676-3

vapor pressure

202 hPa ( 30 °C)

Quality Level

form

liquid

reaction suitability

reagent type: oxidant

pH

1-2 (20 °C in H2O)

mp

11-20 °C

density

1.69 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/2C3F6O.3H2O/c2*4-2(5,6)1(10)3(7,8)9;;;/h;;3*1H2

InChI key

VAIZVCMDJPBJCM-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric, calc. on anhydrous substance): ≥ 97.0 %
Water (K. F.): 14.0 - 18.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service