Skip to Content
Merck
All Photos(1)

Documents

Z680796

Duran® stirred reactor systems

capacity 1000 mL

Synonym(s):

1200396

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

glass bottle
polypropylene cap
round bottle

feature

with cap

packaging

pack of 1 ea

manufacturer/tradename

DURAN® 1200396

capacity

1000 mL

fitting

GL 45 connector (with 2 GL 14 PBT caps, red)
for GL 45 bottles

Looking for similar products? Visit Product Comparison Guide

General description

The stirred reactor is ideal for the widest possible range of mixing processes in laboratories, such as mixing liquids or dissolving solids. The whole assembly is autoclavable, so that it can be used in biological applications, for simple fermentation processes.

  • Drive using standard magnetic stirrers
  • Variable stirrer shaft, can be used for 500 mL and 1,000 mL Duran® GL 45 laboratory bottles
  • Can be used up to 500 rpm
  • Compatible with the Duran® connection systems; hoses with bores from 1.6 mm to 6.0 mm can be fitted

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service